missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BzF

Product Code. 18767866
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767866 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767866 Supplier Tocris Bioscience™ Supplier No. 6786/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Photoactivatable Cross-linking Probes

BzF is a photoreactive unnatural amino acid. Acts as photo cross-linking probe when incorporated into proteins, including ion channels, transporters and GPCRs. When exposed to UV light it forms a covalent cross-link with a C-H bond-containing group, enabling selective and potent UV-driven photoinactivation of protein function. Has been used to study protein-protein and protein-ligand interactions.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Benzoyl-L-phenylalanine
CAS 104504-45-2
Quantity 50 mg
Molecular Formula C16H15NO3
Formula Weight 269.3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.