missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ C 021 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
C 021 dihydrochloride is a potent CCR4 chemokine receptor antagonist (IC50 values are 0.14 and 0.039 μM for inhibition of chemotaxis in human and mouse respectively).
Specifications
Specifications
| CAS | 1784252-84-1 |
| Quantity | 10 mg |
| Product Type | C 021 dihydrochloride |
| Purity | >99% |
| Molecular Weight (g/mol) | 540.57 |
| Inhibitors | C 021 dihydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction