missing translation for 'onlineSavingsMsg'
Learn More

C-276 foil, 0.2mm (0.008in) thick

Product Code. 15421947
Change view
Click to view available options
Quantity:
100 x 100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421947 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15421947 Supplier Thermo Scientific Alfa Aesar Supplier No. 043994.GH
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

High strength, corrosion resistant alloy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material C-276 foil
Name Note 0.2mm (0.008 in.) thick
Quantity 100 x 100 mm
Form Foil
MDL Number MFCD00801065

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.