Learn More
Motic™ C-Mount Camera Adapter for SMZ-160
Description
- C-mount camera adapter for SMZ-160.
- Available in 0.35x, 0.5x, or 1.0x (no lens) magnification.
- Adapt 1/3 in. or 1/2 in. chip sensors.
Specifications
Specifications
| Type | Adapter |
| Autoclavable | Not Autoclavable |
| Camera Type | C-Mount |
| For Use With (Application) | To screw a digital camera on a trinocular SMZ-160 |
| For Use With (Equipment) | To be used with a digital camera for microscopy |
| Magnification Power | 0.35X |
| Warranty | 5 Years |