Learn More
IKA™ C1 Calorimeter Package with C1.12 Combustion Chamber
Product Code. 17267479 Shop All IKA Products
Change view
Click to view available options
Includes:
C1.20 Heater
RC2 Basic Cooling Water Supply
Requires suitable water cooling supply to operate
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
17267479 RC2 Basic Cooling Water Supply Each
17277479 Requires suitable water cooling supply to operate Each
17287479 C1.20 Heater Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17267479 Supplier IKA™ Supplier No. 0010005636
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
The C1 Calorimeter Package with C1.20 Combustion Chamber combines a compact design with a high degree of automation, featuring a light, halogen-resistant combustion chamber.
  • Features a high degree of automation
  • Compact design offers a small footprint
  • Operates in accordance with DIN 51900 and ISO 1928
  • PCI / PCS measurement analysis follows classic isoperibolic methodology
  • Easily retractable combustion chamber with active catalytic coating and halogen-resistance is specially designed for the combustion of sample materials containing halogens and sulfur (>3%)
  • Compatible with IKA™ C6040 Calwin calorimeter software (sold separately), which allows additional extensions and adaptations, such as the management of measurement data and LIMS
  • Includes wear parts and consumables for the first 500 tests including 25 calibrations
  • C1.10 combustion chamber is fitted with a large C5010.5 crucible support and a large C6 quartz crucible
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calorimeter
Certifications/Compliance DIN 51900, ISO 1928
Hertz 50/60 Hz
Includes RC2 Basic Cooling Water Supply
Sample Range Up to 40,000 J
Temperature Range (Metric) 20°C to 30°C
Voltage 100/240 V