missing translation for 'onlineSavingsMsg'
Learn More

C14 Dihydroceramide, TRC

Product Code. 30355207
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30355207 5 mg 5mg
30353307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30355207 Supplier Toronto Research Chemicals Supplier No. TRCC2628305MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61389-70-6
Molecular Formula C32 H65 N O3
Molecular Weight (g/mol) 511.86
IUPAC Name N-[(2S,3R)-1,3-dihydroxyoctadecan-2-yl]tetradecanamide
SMILES CCCCCCCCCCCCCCC[C@@H](O)[C@H](CO)NC(=O)CCCCCCCCCCCCC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C32H65NO3/c1-3-5-7-9-11-13-15-16-18-19-21-23-25-27-31(35)30(29-34)33-32(36)28-26-24-22-20-17-14-12-10-8-6-4-2/h30-31,34-35H,3-29H2,1-2H3,(H,33,36)/t30-,31+/m0/s1
Formula Weight 511.4964
Chemical Name or Material C14 Dihydroceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.