missing translation for 'onlineSavingsMsg'
Learn More

C16-Ceramide, TRC

Product Code. 30364192
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364192 50 mg -
30352913 5 mg 5mg
30362505 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364192 Supplier Toronto Research Chemicals Supplier No. TRCC26290050MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24696-26-2
Molecular Formula C34 H67 N O3
Molecular Weight (g/mol) 537.9
Synonym Hexadecanamide, N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]-, Hexadecanamide, N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]- (9CI), Hexadecanamide, N-[2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]-, [R-[R*,S*-(E)]]-, N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]hexadecanamide, (2S,3R,4E)-2-N-Palmitoyloctadecasphinga-4-ene, C16-Ceramide, C16-Ceramide, C16:0-Ceramide, Ceramide 16, D-erythro-C16-Ceramide, D-erythro-N-Hexadecanoylsphingenine, D-erythro-N-Palmitoylsphingosine, D-erythro-δ4-Ceramide, N-Hexadecanoyl-D-erythro-ceramide, N-Hexadecanoylsphingosine, N-Palmitoyl 4-sphingenine, N-Palmitoyl ceramide, N-Palmitoyl-D-erythro-sphingosine, N-Palmitoyl-D-sphingosine, N-Palmitoylsphingosine
IUPAC Name N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]hexadecanamide
SMILES CCCCCCCCCCCCCCCC(=O)N[C@@H](CO)[C@H](O)\C=C\CCCCCCCCCCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C34H67NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(37)32(31-36)35-34(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h27,29,32-33,36-37H,3-26,28,30-31H2,1-2H3,(H,35,38)/b29-27+/t32-,33+/m0/s1
Formula Weight 537.5121
Percent Purity >95%
Chemical Name or Material C16 Ceramide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.