missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ C19orf80 293T Cell Transient Overexpression Lysate (Denatured)

Product Code. 16121963
Change view
Click to view available options
Quantity: 100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16121963 Quantity: 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16121963 Supplier Abnova™ Supplier No. H00055908T01.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Denatured cell transient overexpression lysate of C19orf80 293T.

  • Transient overexpression cell lysate was tested with Anti-C19orf80 antibody (H00055908-B01) by Western Blots.

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description Gene ID: 55908
Gene Accession No. AAI46553.1
Plasmid pCMV-C19orf80 full-length
Quality Control Testing Transient overexpression cell lysate was tested with Anti-C19orf80 antibody by Western Blots
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Product Type chromosome 19 open reading frame 80
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.