missing translation for 'onlineSavingsMsg'
Learn More

C26 Ceramide, TRC

Product Code. 30012797
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012797 5 mg 5mg
30006952 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012797 Supplier Toronto Research Chemicals Supplier No. TRCC2634555MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121459-09-4
Molecular Formula C44H87NO3
Molecular Weight (g/mol) 678.17
Synonym N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecenyl]hexacosanamide, [R-[R*,S*-(E)]]-N-[2-Hydroxy-1-(hydroxymethyl)-3-heptadecenyl]hexacosanamide;
IUPAC Name N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]hexacosanamide
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO)[C@@H](/C=C/CCCCCCCCCCCCC)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C44H87NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-25-26-28-30-32-34-36-38-40-44(48)45-42(41-46)43(47)39-37-35-33-31-29-27-16-14-12-10-8-6-4-2/h37,39,42-43,46-47H,3-36,38,40-41H2,1-2H3,(H,45,48)/b39-37+/t42-,43+/m0/s1
Chemical Name or Material C26 Ceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.