missing translation for 'onlineSavingsMsg'
Learn More

C26-Dihydro Ceramide, TRC

Product Code. 30379022
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379022 25 mg -
30373564 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379022 Supplier Toronto Research Chemicals Supplier No. TRCD44878025MG
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182362-38-5
Molecular Formula C44H89NO3
Molecular Weight (g/mol) 680.18
Synonym [R-(R*,S*)]-N-[2-Hydroxy-1-(hydroxymethyl)heptadecyl]hexacosanamide, C26-Dihydroceramide, C26-Dihydro Ceramide
IUPAC Name N-[(2S,3R)-1,3-dihydroxyoctadecan-2-yl]hexacosanamide
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO)[C@H](O)CCCCCCCCCCCCCCC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C44H89NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-25-26-28-30-32-34-36-38-40-44(48)45-42(41-46)43(47)39-37-35-33-31-29-27-16-14-12-10-8-6-4-2/h42-43,46-47H,3-41H2,1-2H3,(H,45,48)/t42-,43+/m0/s1
Formula Weight 679.6842
Chemical Name or Material N-[(1S,2R)-2-Hydroxy-1-(hydroxymethyl)heptadecyl]hexacosanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.