Learn More
uvex C300 Wet Cut Protection Gloves
Product Code. p-9625001 Shop All Uvex Products
Change view
Click to view available options
Size:
8
10
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15397082 8 Each
15367082 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15397082 Supplier uvex Supplier No. 6054208
Add to basket
Edge
Add to basket
C300 gloves provide mechanical abrasion resistance with an innovative soft-grip coating. Bamboo TwinFlex™ Technology ensures a high level of cut protection.
  • Soft-grip coating ensures mechanical abrasion resistance in wet applications
  • Bamboo TwinFlex™ Technology provides a high level of cut resistance
  • Highly flexible with good tactile sensitivity
  • Silicone-free according to the imprint test
TRUSTED_SUSTAINABILITY
Specifications
Coated Area Palm and fingertips
Coating Material HPE
Color Green, Black
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Latex Free Yes
Material HPPE, Polyamide, Bamboo Viscose
Product Type Cut-resistant Gloves
Size 8
EN388 Standard 4X42C
For Use With (Application) Automotive industry, assembly, maintenance, metalworking, shipping/logistics, sorting, glass handling, and sheet metal processing
Glove Cuff Style Knit
Model C300 Wet