Learn More
Tocris Bioscience™ C34
GREENER_CHOICE
Product Code. 18795788
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18795788
Brand: Tocris Bioscience™ 5373/10
TLR4 inhibitor
C34 is a TLR4 inhibitor; attenuates TLR4 signaling in enterocytes and macrophages in vitro, and reduces systemic inflammation in mouse models of endotoxemia and necrotizing enterocolitis (NEC). Also suppresses LPS signaling in human ileum displaying NEC ex-vivo. Displays no significant effects on signaling via TLR2 or TLR9.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Product Type C34
Molecular Weight (g/mol) 389.4
Inhibitors C34