missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf80 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
359.00 € - 515.00 €
Specifications
| Antigen | C3orf80 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
C3orf80 Polyclonal antibody specifically detects C3orf80 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| C3orf80 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Chromosome 3 Open Reading Frame 80 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PDDDDDSPALLRDEAAAGSQDSLLDSGG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 401097 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title