missing translation for 'onlineSavingsMsg'
Learn More

uvex C500 Cut Protection Gloves

Product Code. 15327062 Shop All Uvex Products
Change view
Click to view available options
Size:
6
8
9
10
11
Unit Size:
10 pairs
Each
Product Code. Size unitSize
15327062 9 Each
17200431 6 10 pairs
15317062 8 Each
15387052 10 Each
15397052 11 Each
5 options
This item is not returnable. View return policy

Product Code. 15327062

Brand: uvex 6049709

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Uvex C500 gloves combine comfort, flexibility, and tactility with the highest level of cut protection (level 5/C). Soft-grip coating provides mechanical abrasion resistance.

  • Bamboo TwinFlex™ Technology ensures a very high level of cut protection
  • Soft-grip coating provides mechanical abrasion resistance
  • Highly flexible with good tactile sensation
  • Uvex Climazone effectively removes excess warmth and moisture
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Silicone-free according to the imprint test
  • 3D Ergo Technology ensures a secure fit
TRUSTED_SUSTAINABILITY

Specifications

Coating Material Uncoated
Color Green
Hand Ambidextrous
Latex Free Yes
Material Bamboo Viscose, HPPE, Polyamide
Product Type Cut-resistant Gloves
Size 9
EN388 Standard 1X4XC
For Use With (Application) Automotive, Construction, Glass Handling, Logistics, Maintenance, and Sheet Metal Processing
Glove Cuff Style Knit
Series C500
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.