missing translation for 'onlineSavingsMsg'
Learn More

uvex C500 Dry Cut Protection Gloves

Product Code. 17260431 Shop All Uvex Products
Change view
Click to view available options
Applications:
Industrial
Unit Size:
1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Applications Unit Size
17260431 Industrial 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17260431 Supplier uvex Supplier No. 6049906
  • ONLINE EXCLUSIVE PRICE
    17.80 €
    15.90 € / 1 pairs
    Save 1.90 €
    10% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Uvex C500 Dry gloves combine comfort, flexibility, and tactility with the highest level of cut protection (level 5/C). High-performance vinyl (HPV) dots on the palm ensure tactile sensation and enhanced grip capabilites in dry applications.

  • Bamboo TwinFlex™ Technology ensures a very high level of cut protection
  • HPV dots on the palm for enhanced grip capabilities
  • Highly flexible with good tactile sensation
  • Uvex Climazone effectively removes excess warmth and moisture
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Silicone-free according to the imprint test
  • 3D Ergo Technology ensures a secure fit
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material HPV Dots
Color Green, Black
Hand Ambidextrous
Latex Free Yes
Material HPPE, Polyamide, Bamboo
Product Type Cut-resistant Gloves
Size 6
EN388 Standard XX4XC
For Use With (Application) Automotive, Construction, Glass Handling, Logistics, Maintenance, and Sheet Metal Processing
Gender Unisex
Glove Cuff Style Knit
Series C500 Dry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.