missing translation for 'onlineSavingsMsg'
Learn More

C8 Ceramide, TRC

Product Code. 30350644
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30350644 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350644 Supplier Toronto Research Chemicals Supplier No. TRCC2627005MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 74713-59-0
Molecular Formula C26 H51 N O3
Quantity 5 mg
InChI Formula InChI=1S/C26H51NO3/c1-3-5-7-9-10-11-12-13-14-15-16-18-19-21-25(29)24(23-28)27-26(30)22-20-17-8-6-4-2/h19,21,24-25,28-29H,3-18,20,22-23H2,1-2H3,(H,27,30)/b21-19+/t24-,25+/m0/s1
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)CCCCCCC
IUPAC Name N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]octanamide
Molecular Weight (g/mol) 425.69
Formula Weight 425.3869
Chemical Name or Material C8 Ceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.