missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CA 074

Product Code. 30001635
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001635 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001635 Supplier R&D Systems™ Supplier No. 4863/1
  • ONLINE EXCLUSIVE PRICE
    251.68 €
    225.75 € / 1mg
    Save 25.93 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective cathepsin B inhibitor

Inhibitor of cathepsin B (Ki = 2-5 nM). Displays selectivity over cathepsins H and L (Ki = 40-200 μM). Shown to reduce bone metastasis in a 4T1.2 breast cancer model.
TRUSTED_SUSTAINABILITY

Specifications

CAS 134448-10-5
Quantity 1 mg
Product Line Tocris
Molecular Formula C18H29N3O6
Formula Weight 383.44
Purity >98%
Molecular Weight (g/mol) 383.45
Chemical Name or Material CA 074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.