missing translation for 'onlineSavingsMsg'
Learn More

CAB-O-SIL™ M-5, Scintillation Grade

Product Code. 10085520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
150 g
Packaging:
Plastic Bottle
Unit Size:
150g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10085520 150 g Plastic Bottle 150g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10085520 Supplier Thermo Scientific Acros Supplier No. 403731500
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7631-86-9 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography, For fluorescence/luminescence spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

CAS 7631-86-9
Physical Form Crystalline Powder
Density 60 g/l tamped density max.
Color White
Quantity 150 g
Molecular Formula O2Si
Linear Formula SiO2
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Merck Index 15, 8632
Packaging Plastic Bottle
Synonym Silicon dioxide
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
PubChem CID 24261
ChEBI CHEBI:30563
Formula Weight 60.08
Chemical Name or Material CAB-O-SIL™ M-5
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.