Learn More
Cabinet
Product Code. 12953445
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
12953445 Each
1 options
Product Code. 12953445 Supplier Brand Not Defined Supplier No. 88V944G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The range of Viewcab sheet steel storage cabinets are suitable for a number of working environments including laboratories, schools, universities, factories etc. The cabinets are available in three different sizes. Viewcab doors are reinforced and use 6mm laminated safety glass to BS 6206, so you can view your stored items without compromising security. Chrome lever locking handle with semi-concealed internal mechanism. Steel shelves capacity 75kg per shelf UDL. Laminated glass shelves 50kg per shelf UDL.
Specifications
Locking Mechanism 3-way lever locking handle
Material (Shelf) Steel
No. of Doors One
No. of Shelves Two