missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-13 Rat anti-Human, Mouse, Alexa Fluor 647, Clone: 392411, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB3264R-100UG
This item is not returnable.
View return policy
Description
Cadherin-13 Monoclonal specifically detects Cadherin-13 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| Cadherin-13 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P55290 | |
| CDH13 | |
| Mouse myeloma cell line NS0-derived recombinant human Cadherin-13 Glu23-Ala692 Accession # P55290 | |
| RUO | |
| 1012 | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| 392411 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| cadherin 13, H-cadherin (heart), cadherin-13, Cadherin13, CDH13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, T-Cadherin, Truncated cadherin, truncated-cadherin | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human Cadherin-13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) E-Cadherin, rhN-Cadherin, rhP-Cadherin, rhVE-Cadherin, rhCadherin-8, -11, -12 or -17 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction