missing translation for 'onlineSavingsMsg'
Learn More

Cadmium carbonate, pure

Product Code. 15259716 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15259716 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15259716 Supplier Thermo Scientific Acros Supplier No. 212320010
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 513-78-0 | CCdO3 | 172.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 343°C to 412°C
CAS 513-78-0
Quantity 1 kg
Molecular Formula CCdO3
Linear Formula CdCO3
MDL Number MFCD00010918
Fieser 04,67
Merck Index 15,1621
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in diluted acids and in concentrated, solutions of nh4-salts
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES [Cd++].[O-]C([O-])=O
IUPAC Name cadmium(2+);carbonate
Molecular Weight (g/mol) 172.42
PubChem CID 10564
Formula Weight 172.41
Percent Purity 60.0 to 63.0% (Cd)
Grade Pure
Chemical Name or Material Cadmium carbonate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium carbonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H341-Suspected of causing genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.