missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Chloride, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15537894 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15537894 500 g Poly Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15537894 Supplier Avantor J.T.Baker Supplier No. 121201

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 10108-64-2
Physical Form Powder
Quantity 500 g
Molecular Formula CdCl2
MDL Number MFCD00010916
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Cd++]
IUPAC Name dichlorocadmium
Molecular Weight (g/mol) 183.31
PubChem CID 24947
ChEBI CHEBI:35456
Formula Weight 183.31
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 4.07kg
Chemical Name or Material Cadmium Chloride, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.