missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Chloride Hemi(Pentahydrate), 79.5 to 81.0%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15642420 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15642420 100 g Poly Bottle 100g
15662420 500 g Poly Bottle 500g
15652420 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15642420 Supplier Honeywell-Fluka Supplier No. 239208100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 79.5-81.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-78-5
MDL Number MFCD00799071

Specifications

Quantity 100 g
Linear Formula CdCl2·21/2H2O
UN Number UN2570
Formula Weight 228.36g/mol
Percent Purity 79.5 to 81.0%
Grade ACS Reagent
Packaging Poly Bottle
Vapor Pressure 10mmHg (656°C)
Density 3.327g/cm3; Vapor Density: 6.3 (vs air)
Chemical Name or Material Cadmium Chloride Hemi(Pentahydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.