missing translation for 'onlineSavingsMsg'
Learn More

Cadmium molybdenum oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11339347
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339347 250 g 250g
11329347 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339347 Supplier Thermo Scientific Alfa Aesar Supplier No. 040202.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Electronic and optical applications

Electronic and optical applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13972-68-4
Molecular Formula CdMoO4
Molecular Weight (g/mol) 272.36
MDL Number MFCD00015997
InChI Key SQHWLBOPUKZGNS-UHFFFAOYSA-N
Synonym cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum
PubChem CID 22420716
IUPAC Name cadmium(2+);dioxido(dioxo)molybdenum
SMILES [O--].[O--].[O--].[O--].[Mo].[Cd++]

Specifications

Melting Point ∼1250°C
Physical Form Powder
Quantity 250 g
UN Number UN2570
Solubility Information Slightly soluble in water; Soluble in acid
Formula Weight 272.35
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Cadmium molybdenum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.