missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, 99+%

Product Code. 10066113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10066113 1 kg Glass bottle 1kg
10511851 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10066113 Supplier Thermo Scientific Acros Supplier No. 212340010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Color Colorless or White
Physical Form Crystals or Powder
Quantity 1 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula Cd(NO3)2·4H2O
Merck Index 15, 1627
Solubility Information Solubility in water: 2150g/L (20°C). Other solubilities: soluble in ammonia,practically insoluble in conc. h2so4,soluble in alcohol,acetone and ethyl acetate
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
Formula Weight 308.46
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Cadmium nitrate tetrahydrate, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium nitrate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Oxidising solid Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.