missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Nitrate Tetrahydrate, purum p.a., ≥99.0% (T), Honeywell™ Riedel-de-Haën™

Product Code. 15681880 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15681880 500 g 500g
15671880 25 g 25g
15661880 500 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15681880 Supplier Honeywell Research Chemicals Supplier No. 20911500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+) bis(nitric acid)
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Quantity 500 g
Linear Formula Cd(NO3)2 · 4H2O
UN Number UN2570
Beilstein 8156730
Formula Weight 308.48g/mol
Percent Purity ≥99.0% (T)
Grade purum p.a.
Chemical Name or Material Cadmium Nitrate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.