missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals

Product Code. 10616431 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10616431

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-84-3 | Cd3O12S3·8H2O | 769.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
Molecular Formula Cd3O12S3·8H2O
Molecular Weight (g/mol) 769.51
MDL Number MFCD02253000

Specifications

Melting Point 41.0°C
Color White
Physical Form Crystals
Quantity 500 g
Linear Formula 3CdSO4·8H2O
Merck Index 15,1632
Solubility Information Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol, acids, acetone and ammonia
Formula Weight 769.51
Percent Purity 98.0 to 102.0%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium sulfate, 8/3-hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.