missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulphate, TRC

Product Code. 30329169
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30329169 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30329169

Brand: Toronto Research Chemicals TRCC0590205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cadmium Sulphate
CAS 10124-36-4
Molecular Formula CdSO4
Quantity 5 g
Synonym Cadmium Monosulfate, Cadmium Sulfate (Cd(SO4)), Cadmium Sulforicum
InChI Formula InChI=1S/Cd.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
SMILES [Cd+2].[O-]S(=O)(=O)[O-]
IUPAC Name cadmium(2+);sulfate
Molecular Weight (g/mol) 208.47
Formula Weight 209.8551
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.