missing translation for 'onlineSavingsMsg'
Learn More

Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15422277
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15422277 25 g Glass bottle 25g
15402277 1 g Plastic bottle 1g
15412277 5 g Plastic bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422277 Supplier Thermo Scientific Alfa Aesar Supplier No. 045457.14
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Used to make thin film solar cells.

Cadmium telluride is used to make thin film solar cells. It can be alloyed with mercury to make a versatile infrared detector material (HgCdTe). It is used as an infrared optical material for optical windows and lenses and is proven to provide a good performance across a wide range of temperatures. It is also applied for electro-optic modulators and has the greatest electro-optic coefficient of the linear electro-optic effect among II-VI compound crystals. It can be doped with chlorine, which is used as a radiation detector for x-rays, gamma rays, beta particles and alpha particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-25-8
Molecular Formula CdTe
Molecular Weight (g/mol) 240.01
MDL Number MFCD00015998
InChI Key RPPBZEBXAAZZJH-UHFFFAOYSA-N
Synonym cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
PubChem CID 91501
IUPAC Name tellanylidenecadmium
SMILES [Cd]=[Te]

Specifications

Melting Point 1041°C
Physical Form Polycrystalline Pieces
Quantity 25 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 240
Percent Purity 99.999%
Packaging Glass bottle
Specific Gravity 6.2
Density 6.2
Chemical Name or Material Cadmium telluride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.