missing translation for 'onlineSavingsMsg'
Learn More

Caffeic acid, MedChemExpress

Product Code. 16469016
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469016 100 mg 100mg
16441328 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469016 Supplier Medchem Express Supplier No. HYN0172

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress Caffeic acid is an inhibitor of both TRPV1 ion channel and 5-Lipoxygenase (5-LO).

Caffeic acid is an inhibitor of both TRPV1 ion channel and 5-Lipoxygenase (5-LO).

Specifications

Chemical Name or Material Caffeic acid
CAS 331-39-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C9H8O4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (555.06 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.55 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 98.36%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.