missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeic Acid Phenethyl Ester, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Caffeic Acid Phenethyl Ester (CAPE) |
| CAS | 104594-70-9 |
| Molecular Formula | C17 H16 O4 |
| Quantity | 25 mg |
| Synonym | 2-Phenylethyl 3-(3,4-dihydroxyphenyl)prop-2-enoate, 2-Phenylethyl caffeate, CAPE, Caffeic acid phenethyl ester, β-Phenylethyl caffeate, Caffeic Acid Phenylethyl Ester |
| InChI Formula | InChI=1S/C17H16O4/c18-15-8-6-14(12-16(15)19)7-9-17(20)21-11-10-13-4-2-1-3-5-13/h1-9,12,18-19H,10-11H2/b9-7+ |
| SMILES | Oc1ccc(\C=C\C(=O)OCCc2ccccc2)cc1O |
| IUPAC Name | 2-phenylethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
| Molecular Weight (g/mol) | 284.31 |
| Formula Weight | 284.1049 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction