missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, 1mg/ml in methanol

Product Code. 10754204 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10754204 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10754204 Supplier Thermo Scientific Acros Supplier No. 393540010
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 58-08-2
Density 0.7910g/mL
Flash Point 12°C
Packaging Glass bottle
Molecular Formula C8H10N4O2
MDL Number MFCD00005758
Quantity 1 mL
Merck Index 15, 1639
Specific Gravity 0.791
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
Molecular Weight (g/mol) 194.19
PubChem CID 2519
ChEBI CHEBI:27732
Formula Weight 194.19
Chemical Name or Material Caffeine, 1mg/mL in methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Caffeine, 1mg/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.