missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Caffeine

Product Code. 18712544
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18712544 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18712544 Supplier Tocris Bioscience™ Supplier No. 2793/100
  • ONLINE EXCLUSIVE PRICE
    96.00 €
    90.30 € / 100mg
    Save 5.70 €
    5% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A1 and A2B antagonist. CNS stimulant

Central nervous system stimulant. Antagonist at A1 and A2A adenosine receptors and inhibitor of cyclic nucleotide phosphodiesterases. Mobilizes calcium from intracellular stores and inhibits benzodiazepine binding to GABA receptors.
TRUSTED_SUSTAINABILITY

Specifications

CAS 58-08-2
Quantity 100 mg
Molecular Formula C8H10N4O2
Formula Weight 194.19
Purity >99%
Molecular Weight (g/mol) 194.19
Chemical Name or Material Caffeine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.