missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Ion Selective Electrode

Product Code. 11735648 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Parameter:
Calcium
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Parameter Unit Size
11735648 Calcium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11735648
  • ONLINE EXCLUSIVE PRICE
    513.00 €
    323.35 € / Pack of 1
    Save 189.65 €
    36% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Ion selective combination pH electrode with a reproducibility better than 2%.

  • Reproducibility better than 2%
  • Interchangeable membrane
  • S7 compliant per-head connection
  • All pH/mV meters or ionometers
  • pH combination gamma 3 to 10

Specifications

pH Range 3 to 10
For Use With (Equipment) pH/mV Meters, Ionometers
Type Ion Selective pH Electrode
Parameter Calcium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.