missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ CalcCheck standard for HI-96710
Click to view available options
For Use With (Equipment):
HI-96710
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | HI-96710 |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P391-Collect spillage.
- MIXTURE LIST-Contains : Hydrochloric acid, cobalt (II) chloridehexahydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction