missing translation for 'onlineSavingsMsg'
Learn More

Calcitriol Impurities D, MedChemExpress

Product Code. 16471979
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471979 1 mg 1mg
16430668 5 mg 5mg
16428228 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471979 Supplier Medchem Express Supplier No. HY77274
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Calcitriol Impurities D is the impurity of Calcitriol, Calcitriol is the hormonally active form of vitamin D, Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).

Calcitriol Impurities D is the impurity of Calcitriol, Calcitriol is the hormonally active form of vitamin D, Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).

Specifications

Chemical Name or Material Calcitriol Impurities D
CAS 103656-40-2
Color White
Purity Grade Notes Research
Molecular Formula C28H46O3
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (232.20 mM; Need ultrasonic)
SMILES O[C@H](C/1=C)C[C@H](O)CC1=C\C=C2[C@@](CC[C@]3([H])[C@@H](CCCCC(C)(O)C)C)([H])[C@]3(C)CCC\2
Molecular Weight (g/mol) 430.66
Formula Weight 430.66
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.