missing translation for 'onlineSavingsMsg'
Learn More

Calcitroic Acid, TRC

Product Code. 30134752
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134752 5 mg 5mg
30138716 10 mg 10mg
30012086 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134752 Supplier Toronto Research Chemicals Supplier No. TRCC1445505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71204-89-2
Molecular Formula C23 H34 O4
Molecular Weight (g/mol) 374.51
Synonym 1H-Indene-1-propanoic acid, 4-[(2Z)-2-[(3S,5R)-3,5-dihydroxy-2-methylenecyclohexylidene]ethylidene]octahydro-β,7a-dimethyl-, (βR,1R,3aS,4E,7aR)-, 24-Nor-9,10-secochola-5,7,10(19)-trien-23-oic acid, 1,3-dihydroxy-, (1α,3β,5Z,7E)- (9CI), (βR,1R,3aS,4E,7aR)-4-[(2Z)-2-[(3S,5R)-3,5-Dihydroxy-2-methylenecyclohexylidene]ethylidene]octahydro-β,7a-dimethyl-1H-indene-1-propanoic acid, 1α-Hydroxycalcioic acid, Calcitroic acid, MC 1235
IUPAC Name (3R)-3-[(1R,3aS,4E,7aR)-4-[(2Z)-2-[(3S,5R)-3,5-dihydroxy-2-methylidenecyclohexylidene]ethylidene]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-1-yl]butanoic acid
SMILES C[C@H](CC(=O)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H34O4/c1-14(11-22(26)27)19-8-9-20-16(5-4-10-23(19,20)3)6-7-17-12-18(24)13-21(25)15(17)2/h6-7,14,18-21,24-25H,2,4-5,8-13H2,1,3H3,(H,26,27)/b16-6+,17-7-/t14-,18-,19-,20+,21+,23-/m1/s1
Formula Weight 374.2457
Chemical Name or Material Calcitroic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.