missing translation for 'onlineSavingsMsg'
Learn More

Calcium, 99%, pieces, Thermo Scientific Chemicals

Product Code. 10053953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10053953 2.5 kg Plastic drum 2.5kg
10579253 5 g Plastic Bottle 5g
10338580 100 g Plastic Bottle 100g
10226450 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10053953 Supplier Thermo Scientific Acros Supplier No. 201180025
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym ingot, elemental, calcium, elemental, atom, kalzium, sy7q814vup, compounds, chelate, hsdb 273, unii-sy7q814vup
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Gray
Melting Point 845.0°C
Boiling Point 1484.0°C
Quantity 2.5 kg
Packaging Plastic drum
Assay Percent Range 99%
Form Granular
Linear Formula Ca
Fieser 05,89; 09,1634; 14,71
Merck Index 15, 1648
Formula Weight 40.07
Percent Purity 99%
Chemical Name or Material Calcium, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Calcium
Signal Word
  • Danger
Hazard Category
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P223-Do not allow contact with water.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P370+P378-In case of fire: Use carbon dioxide to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.