missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, 99%

Product Code. 11497127
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11497127

Brand: Thermo Scientific Alfa Aesar A14947.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol

Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Density 1.5
Linear Formula Ca(OOCCH3)2·xH2O
Quantity 100 g
Beilstein 3692527
Merck Index 14,1646
Solubility Information Soluble in water.
Formula Weight 158.17 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Calcium acetate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.