Learn More
Calcium acetate monohydrate, ACS, 99.0% min
Description
Calcium acetate monohydrate is used to precipitate oxalates in solution. It is used to control the phosphate levels in blood, thereby controlling the bone problems. It is also involved in the neutralization of fluoride in water. Further, it is used as a food additive mainly in candy products. In addition to this, it serves as a stabilizer, buffer and sequestrant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | White |
| Physical Form | Powder |
| Quantity | 250 g |
| Sensitivity | Hygroscopic |
| Assay Percent Range | 99% |
| Density | 1.5 g/cm3 |
| Beilstein | 3692527 |
| Solubility Information | Soluble in water. Slightly soluble in methanol and hydrazine. Insoluble in ethanol,acetone and benzene. |
| Formula Weight | 176.18 (158.17 Anhydrous) |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.