missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate Monohydrate, ACS Reagent, ≥99%, Solstice

Product Code. 15605410
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15605410 500 g Poly Bottle 500g
15695400 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15605410 Supplier Honeywell-Fluka Supplier No. 402850500G
  • ONLINE EXCLUSIVE PRICE
    302.00 €
    272.58 € / 500g
    Save 29.42 €
    9% Off
View all promotions
 Restricted Item
Request Bulk or Custom Format
Edge
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.181
MDL Number MFCD00150019
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
PubChem CID 82163
ChEBI CHEBI:59199
IUPAC Name calcium;diacetate;hydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

Specifications

Color White
Quantity 500 g
Linear Formula Ca(CH3CO2)2 · H2O
UN Number NONH for all modes of transport
Formula Weight 176.18g/mol
Percent Purity ≥99%
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Calcium acetate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.