missing translation for 'onlineSavingsMsg'
Learn More

Calcium benzoate hydrate, 98%, water <8%

Product Code. 11439493
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439493 100 g 100g
11449493 500 g 500g
11459493 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439493 Supplier Thermo Scientific Alfa Aesar Supplier No. A12358.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2090-05-3 | C19H20O4 | 312.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Calcium benzoate hydrate is used in used in the food industry as a preservative.

Solubility
Soluble in water (2.6 g/100ml at 0°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2090-05-3

Specifications

Molecular Formula C19H20O4
MDL Number MFCD00167164
Quantity 100 g
Synonym bis 4-glycidyloxyphenyl methane, bis 4-glycidyloxy phenyl methane, bis 4-oxiran-2-yl methoxy phenyl methane, para-para-bfdge, p,p-bfdge, diphenylolmethane diglycidyl ether, 2,2'-methylenebis p-phenyleneoxymethylene bisoxirane, bis p-2,3-epoxypropoxy phenyl methane, 4,4'-methylenebisphenol diglycidyl ether, oxirane, 2,2'-methylenebis 4,1-phenyleneoxymethylene bis
Solubility Information Soluble in water (2.6g/100ml at 0°C).
InChI Key XUCHXOAWJMEFLF-UHFFFAOYNA-N
SMILES C1C(O1)COC2=CC=C(C=C2)CC3=CC=C(C=C3)OCC4CO4
IUPAC Name 2-[[4-[[4-(oxiran-2-ylmethoxy)phenyl]methyl]phenoxy]methyl]oxirane
Molecular Weight (g/mol) 312.37
PubChem CID 91511
Formula Weight 282.32 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Calcium benzoate hydrate, Water <8%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.