missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide hydrate, for analysis

Product Code. 10572271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10572271 1 kg Plastic bottle 1kg
10480911 500 g Plastic bottle 500g
10544664 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10572271 Supplier Thermo Scientific Acros Supplier No. 300840010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 71626-99-8 | Br2Ca | 199.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]

Specifications

Melting Point 730.0°C
Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 1 kg
Linear Formula CaBr2·xH2O
Solubility Information Solubility in water: 1420g/L (20°C). Other solubilities: soluble in methanol, ethanol
Formula Weight 199.9
Percent Purity ≥83.0% (Complexometry) (CaBr2)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium bromide hydrate, For analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.