missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbide, 72-82%

Product Code. 10762284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762284 1 kg Glass bottle 1kg
10646124 250 g Glass bottle 250g
10219103 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10762284 Supplier Thermo Scientific Acros Supplier No. 389790010
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-20-7 | C2Ca | 64.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-20-7
Molecular Formula C2Ca
Molecular Weight (g/mol) 64.1
MDL Number MFCD00010905
Synonym calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate

Specifications

Boiling Point 2300.0°C
Melting Point 2160.0°C
Color Gray
Physical Form Lump
Quantity 1 kg
Assay Percent Range 12 to 23% (CaO) (Complexometry)
Linear Formula CaC2
Merck Index 15, 1658
Solubility Information Solubility in water: hydrolyses
Formula Weight 64.1
Percent Purity 72 to 82%
Identification Positive
Packaging Glass bottle
Chemical Name or Material Calcium carbide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.