missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbide, Technical, Fisher Chemical

Product Code. 10253850 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10253850 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10253850 Supplier Fisher Chemical Supplier No. C/1000/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 2300°C
Melting Point 2160°C
CAS 75-20-7
Color Gray
Physical Form Solid
Quantity 500 g
Molecular Formula C2Ca
MDL Number 10905
InChI Key UIXRSLJINYRGFQ-UHFFFAOYSA-N
SMILES [Ca++].[C-]#[C-]
IUPAC Name calcium acetylene
Molecular Weight (g/mol) 64.10
Formula Weight 64.1g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Calcium carbide
Signal Word
  • Danger
Hazard Category
  • Substance which, in contact with water, emits flammable gases Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P231-Handle under inert gas.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Calcium carbide, Calcium oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.