missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, chelometric standard, ACS, 99.95-100.05%

Product Code. 11387858
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387858 100 g 100g
11397858 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387858 Supplier Thermo Scientific Alfa Aesar Supplier No. 036399.22
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calcium carbonate, CCaO3, CAS Number-471-34-1, aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble, 500g, White powder, 99.95-100.05%, CHEBI:3311, 2.93, 207-439-9, 100.09, ACS, VTYYLEPIZMXCLO-UHFFFAOYSA-L, calcium;carbonate | CAS: 471-34-1 | CCaO3 | 100.09 g/mol

Calcium carbonate is a dietary supplement used when the quantity of calcium taken in the diet is not enough. Used as an antacid to relieve heartburn, acid indigestion and upset belly. Practiced in the expression industry. Also widely used as a filler in plastics and is used in metallurgy of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.95-100.05%
Solubility Information Insoluble in water but soluble in acids.
IUPAC Name calcium carbonate
Formula Weight 100.09
Grade ACS
Refractive Index 1.6583
Appearance White powder
Density 2.93 g/cm3 at 20°C
Chemical Name or Material Calcium carbonate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.