missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 1M aq. soln.

Product Code. 15445389
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445389 50 mL 50mL
15455389 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445389 Supplier Thermo Scientific Alfa Aesar Supplier No. J63122.AD
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol

Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong acids, borane/boron oxides, zinc, calcium oxide, methyl vinyl ether. Calcium chloride is attacked by bromine trifluoride. Anhydrous Calcium Chloride is highly hygroscopic and is often used as a desiccant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Solubility Information Soluble in water (74.5g/100ml) at 20°C,alcohol,water (159g/100ml) at 100°C,acetic acid,and acetone.
Formula Weight 110.99
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Odor Odorless
Chemical Name or Material Calcium chloride
compliance-icons
Product Identifier
  • Calcium chloride, 1M aq. soln
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: calcium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.