missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, Dihydrate, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Description
- Calcium chloride is a commonly used reagent in biochemistry.
- It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.
- Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli.
- It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.
- Calcium Chloride is a salt of calcium and chloride.
- It behaves as a typical ionic halide, and is solid at room temperature
Specifications
Specifications
| Melting Point | 176°C (Decomposition)(Literature) |
| CAS | 10035-04-8 |
| pH | 4 to 9.5 (5% aq soln) |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | ∽99% |
| Molecular Formula | CaCl2·2H2O |
| MDL Number | MFCD00149613 |
| Synonym | Calcium dichloride dihydrate |
| Solubility Information | Soluble in water - 294mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction