missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate, MP Biomedicals™

Product Code. 11465195 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465195 500 g 500g
11475195 1 kg 1kg
11455655 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465195 Supplier MP Biomedicals Supplier No. 02199524.5
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Useful in preparing competent bacteria. Calcium Chloride Dihydrate, MP Biomedicals is a USP-grade biochemical reagent. It is used in the preparation of calcium-alginate beads and to compose the medium for Thermophilic Clostridia (MTC). It may be used for

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2. 2H2O

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water with heat liberation, ethanol, acetic acid, acetone.
Formula Weight 147.01
Percent Purity 99.0 to 107.0%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.