missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, puriss., Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
2.5 kg
15 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
15kg
1kg
2.5kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 6 x 2.5 kg |
| UN Number | NONH for all modes of transport |
| Formula Weight | 147.01 |
| Percent Purity | 99 to 103% |
| Grade | puriss. |
| Chemical Name or Material | Calcium Chloride Dihydrate, Meets analytical specification of Ph. Eur.USP, FCC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction